Learn More
MilliporeSigma™ TAE Buffer, 10X, Molecular Biology Grade, Calbiochem™,
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Chemical Name or Material | TAE Buffer |
| CAS | 77-86-1, 7732-18-5 |
| For Use With (Application) | Agarose Gel Electrophoresis, Polyacrylamide Gel Electrophoresis |
| pH | 8.3 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Solution Type | Buffer |
| DNase | None Detected |
| Protease | None Detected |
| Synonym | Tris-Acetate-EDTA Buffer |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.