Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAE Buffer, 10X Solution, Molecular Biology Grade, Ultrapure

Catalog No. AAJ75904K2 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
5 L
Packaging:
Nalgene bottle
Upright carton
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ75904K2 1 L Nalgene bottle
AAJ75904K8 5 L Upright carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ75904K2 Supplier Thermo Scientific Supplier No. J75904K2
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material TAE Buffer
Concentration 10X
Physical Form Solution
Quantity 1 L
Packaging Nalgene bottle
Grade Molecular Biology, Ultrapure
What are the concentrations of the components of TAE Buffer, 10X Solution, Molecular Biology Grade, Ultrapure (Cat. No. J75904.K8)?

TAE Buffer, 10X Solution, Molecular Biology Grade, Ultrapure contains 400 mM Tris and 0.01 M EDTA in high-purity RO water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.