Learn More
Growcells TAE BUFFER 50X 1L EA
Catalog No. 501034829
Encompass_Preferred
Sterile liquid electrophoresis buffer concentrate prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging. Recipe/L: 2424.8g TRIS Base, 570ml Acetic Acid, 186.1g EDTA Ph 8.3
Catalog No. 50-103-4829 Supplier Growcells Supplier No. MRGF4210
Add to Cart
Add to Cart
Sterile liquid electrophoresis buffer concentrate prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging. Recipe/L: 2424.8g TRIS Base, 570ml Acetic Acid, 186.1g EDTA Ph 8.3