Learn More
Growcells TAE BUFFER 50X 2L EA
Catalog No. 501034831
Encompass_Preferred
Sterile liquid electrophoresis buffer concentrate prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging. Recipe/L: 2424.8g TRIS Base, 570ml Acetic Acid, 186.1g EDTA Ph 8.3
Catalog No. 50-103-4831 Supplier Growcells Supplier No. MRGF4211
Sterile liquid electrophoresis buffer concentrate prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging. Recipe/L: 2424.8g TRIS Base, 570ml Acetic Acid, 186.1g EDTA Ph 8.3