Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAE Buffer (Tris-Acetate-EDTA Buffer) Concentrate, Innovating Science™
SCI_ED

Catalog No. S02446 Shop All Aldon Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02446 100 mL
S02408 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S02446 Supplier Aldon Corporation Supplier No. IS29032
Add to Cart
Add to Cart

Specifications

CAS 7732-18-5
For Use With (Application) Molecular Biology
For Use With (Equipment) Mini Precast Protein Gels
Physical Form Liquid
Quantity 100 mL
Solution Type Buffer
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.