Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TAE Powder / TAE Buffer | 100 POUCHES
SDP

Catalog No. 5000430188
Encompass_Preferred

TAE (Tris-acetate-EDTA) buffer is widely used in biological experiments, especially in agarose gel electrophoresis of nucleic acid. Linear dsDNA migrates quickly in electrophoresis, and DNA fragments (> 13 kb) can be separated effectively. TAE buffer is also suitable for DNA gel extraction.

  • Consists of Tris-acetate and EDTA-2Na.
  • Each pouch can be diluted to 1 L of 1x TAE buffer.
  • Concentration of Tris-acetate is 40 mM and EDTA-2Na is 1 mM when diluted to 1 L of 1x TAE buffer.
  • Provides a convenient way to make TAE solution.
  • Eliminates the need to weigh and mix individual components.
  • pH of the 1x TAE buffer is 8.3 ± 0.1 @ 25°C.

Catalog No. 50-004-30188 Supplier Medchemexpress LLC Supplier No. HYK1015100POUCHES
May include imposed supplier surcharges.
Add to Cart
Add to Cart

TAE (Tris-acetate-EDTA) buffer is widely used in biological experiments, especially in agarose gel electrophoresis of nucleic acid. Linear dsDNA migrates quickly in electrophoresis, and DNA fragments (> 13 kb) can be separated effectively. TAE buffer is also suitable for DNA gel extraction.

  • Consists of Tris-acetate and EDTA-2Na.
  • Each pouch can be diluted to 1 L of 1x TAE buffer.
  • Concentration of Tris-acetate is 40 mM and EDTA-2Na is 1 mM when diluted to 1 L of 1x TAE buffer.
  • Provides a convenient way to make TAE solution.
  • Eliminates the need to weigh and mix individual components.
  • pH of the 1x TAE buffer is 8.3 ± 0.1 @ 25°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.