Learn More
Kd Medical Inc TAE (TRIS-ACETATE EDTA), 50X
SDP
Catalog No. 501031380
Encompass_Preferred
TAE Buffer is the most commonly used buffer for agarose DNA electrophoresis. 50X concentrated ready to be diluted to a 1X working solution. One 500ml bottle.
Catalog No. 50-103-1380 Supplier Kd Medical Inc Supplier No. RGE3320
TAE Buffer is the most commonly used buffer for agarose DNA electrophoresis. 50X concentrated ready to be diluted to a 1X working solution. One 500ml bottle.