Learn More
Selleck Chemical LLC TAE684 (NVP-TAE684) 5mg 761439-42-3
SDP
Catalog No. 508831
Encompass_Preferred
TAE684 (NVP-TAE684) is a potent and selective ALK inhibitor with IC50 of 3 nM in a cell-free assay, 100-fold more sensitive for ALK than InsR. TAE684 (NVP-TAE684) induces cell cycle arrest and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-883-1 Supplier Selleck Chemical LLC Supplier No. S11085MG
TAE684 (NVP-TAE684) is a potent and selective ALK inhibitor with IC50 of 3 nM in a cell-free assay, 100-fold more sensitive for ALK than InsR. TAE684 (NVP-TAE684) induces cell cycle arrest and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.