Learn More
Selleck Chemical LLC Tenofovir alafenamide hemifumarate S4427-5mg
SDP
Catalog No. 503110194
Encompass_Preferred
Tenofovir alafenamide (TAF GS-7340) hemifumarate is a prodrug of tenofovir but results in significantly higher intracellular tenofovir concentrations and lower serum levels Tenofovir alafenamide is a novel nucleotide reverse transcriptase inhibitor for the treatment of HIV-1 infection
Catalog No. 50-311-0194 Supplier Selleck Chemical LLC Supplier No. S44275MG
Tenofovir alafenamide (TAF GS-7340) hemifumarate is a prodrug of tenofovir but results in significantly higher intracellular tenofovir concentrations and lower serum levels Tenofovir alafenamide is a novel nucleotide reverse transcriptase inhibitor for the treatment of HIV-1 infection