Learn More
Aviva Systems Biology Corp TAF1 Antibody - middle region (ARP32472_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32472_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TAF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TAF1. Alternate names: BA2R, CCG1, CCGS, DYT3, KAT4, NSCL2, OF, P250, TAF2A, TAFII250, XDP, N-TAF1, DYT3/TAF1ApplicationWB, CHIP. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54610