Learn More
Aviva Systems Biology Corp TAF10 Antibody - middle region (ARP31425_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31425_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TAF10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TAF10. Alternate names: TAF2A, TAF2H, TAFII30ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54214