Learn More
Aviva Systems Biology Corp TAF11 Antibody - N-terminal region (ARP32136_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32136_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TAF11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TAF11. Alternate names: TAF2I, PRO2134, TAFII28, MGC:15243ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54460