Learn More
Aviva Systems Biology Corp TAF13 Antibody - N-terminal region (ARP47596_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47596_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TAF13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TAF13. Alternate names: MGC22425, TAF2K, TAFII18, TAFII-18, TAF(II)18ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59602