Learn More
Aviva Systems Biology Corp TAF15 Antibody - N-terminal region (ARP30112_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30112_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TAF15. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TAF15. Alternate names: Npl3, RBP56, TAF2N, TAFII68, hTAFII68ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-54131