Learn More
Aviva Systems Biology Corp TAF1A Antibody - C-terminal region (P100802_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100802_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TAF1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TAF1A. Alternate names: SL1, RAFI48, TAFI48, MGC:17061ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54089