Learn More
Aviva Systems Biology Corp Taf1b Antibody - middle region (ARP37290_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37290_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Taf1b. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of mouse Taf1b. Alternate names: 4930408G01Rik, A230108M10Rik, Tafi86, mTAFI68, TAFI68ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-613