Learn More
Aviva Systems Biology Corp Taf1c Antibody - C-terminal region (ARP40130_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40130_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Taf1c. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Tafi95, mTAFI95ApplicationWB. Species reactivity: Guinea Pig, Mouse, Rat
Catalog No. 50-284-853