Learn More
Aviva Systems Biology Corp TAF4 Antibody - middle region (ARP33394_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33394_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TAF4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TAF4. Alternate names: FLJ41943, TAF2C, TAF2C1, TAF4A, TAFII130, TAFII135ApplicationWB, CHIP. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit
Catalog No. 50-014-54996