Learn More
Aviva Systems Biology Corp TAF5L Antibody - N-terminal region (ARP39081_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39081_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TAF5L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TAF5L. Alternate names: PAF65BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56905