Learn More
Aviva Systems Biology Corp TAF6 Antibody - N-terminal region (ARP34693_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34693_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TAF6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TAF6. Alternate names: DKFZp781E21155, MGC:8964, TAF2E, TAFII70, TAFII80, TAFII85, TAFII-70, TAFII-80, TAF(II)70, TAF(II)80ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-014-55515