Learn More
Aviva Systems Biology Corp TAF6 Antibody - C-terminal region (ARP38653_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38653_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TAF6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TAF6. Alternate names: DKFZp781E21155, MGC:8964, TAF2E, TAFII70, TAFII80, TAFII85, TAFII-70, TAFII-80, TAF(II)70, TAF(II)80ApplicationWB, CHIP. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56742