Learn More
Aviva Systems Biology Corp TAF6L Antibody - N-terminal region (ARP38801_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38801_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TAF6L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TAF6L. Alternate names: PAF65AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56806