Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci TAF9 Peptide
SDP

Catalog No. 16596354
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-596-354 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-596-354 Supplier ProSci Supplier No. 7719P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

The peptide is used for blocking the activity of TAF9 antibody (catalog number 7719). It usually blocks the antibody activity completely by incubating the peptide with equal volume of antibody for 30 min at 37°C.

Specifications

Gene ID (Entrez) 6880
Species Human
Accession Number NP_003178
Common Name TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 32 kDa
Concentration 200 μg/mL
Content And Storage Store TAF9 peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol TAF9
Quantity 0.05 mg
Sequence 17 amino acid peptide near the center of human TAF9.
Synonym TAF2G, TAFII31, TAFII32, MGC:5067, TAFII-31, TAFII-32, TAFIID32, STAF31/32, TAF2G, Transcription initiation factor TFIID subunit 9, RNA polymerase II TBP-associated factor subunit G
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.