Learn More
Medchemexpress LLC HY-14852 5mg Medchemexpress, Tafamidis CAS:594839-88-0 Purity:>98%
SDP
Catalog No. 502081649
Encompass_Preferred
Medchemexpress, HY-14852 5mg Tafamidis CAS:594839-88-0 Tafamidis is a potent and selective transthyretin (TTR) stabilizer, shows comparable potency and efficacy to the mutumant homotetramers V30M-TTR, V122I-TTR and wild type WT-TTR, with EC50s of 2.7-3.2 μM. Tafamidis inhibits amyloidogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1649 Supplier Medchemexpress LLC Supplier No. HY148525MG
Medchemexpress, HY-14852 5mg Tafamidis CAS:594839-88-0 Tafamidis is a potent and selective transthyretin (TTR) stabilizer, shows comparable potency and efficacy to the mutumant homotetramers V30M-TTR, V122I-TTR and wild type WT-TTR, with EC50s of 2.7-3.2 μM. Tafamidis inhibits amyloidogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.