Learn More
Medchemexpress LLC Tafamidis meglumine | 951395-08-7 | MFCD28502032 | 99.8% | 503.33 g/mol | C21H24Cl2N2O8 | 10 MG
SDP
Catalog No. 5000287142
Encompass_Preferred
Tafamidis meglumine is the meglumine salt form of tafamidis, a potent and selective transthyretin (TTR) stabilizer used in research to inhibit TTR amyloidogenesis. Supplied as a solid, high-purity compound, it is intended for in vitro and biochemical studies of TTR stabilization and amyloid diseases.
  • Potent and selective transthyretin stabilizer.
  • Inhibits transthyretin amyloidogenesis in biochemical assays.
  • High purity (99.8%).
  • Meglumine salt supplied as a solid.
  • Suitable for in vitro and preclinical research applications.
  • Available in small packaging for laboratory-scale studies.
Catalog No. 50-002-87142 Supplier Medchemexpress LLC Supplier No. HY14852A10MG
Tafamidis meglumine is the meglumine salt form of tafamidis, a potent and selective transthyretin (TTR) stabilizer used in research to inhibit TTR amyloidogenesis. Supplied as a solid, high-purity compound, it is intended for in vitro and biochemical studies of TTR stabilization and amyloid diseases.
  • Potent and selective transthyretin stabilizer.
  • Inhibits transthyretin amyloidogenesis in biochemical assays.
  • High purity (99.8%).
  • Meglumine salt supplied as a solid.
  • Suitable for in vitro and preclinical research applications.
  • Available in small packaging for laboratory-scale studies.