Learn More
Selleck Chemical LLC Tafenoquine E2665-25MG
SDP
Catalog No. 5000040484
Encompass_Preferred
Tafenoquine (WR 238605) is an 8-aminoquinoline used as an anti-malarial prophylactic agent
Catalog No. 50-000-40484 Supplier Selleck Chemical LLC Supplier No. E266525MG
Tafenoquine (WR 238605) is an 8-aminoquinoline used as an anti-malarial prophylactic agent