Learn More
Selleck Chemical LLC Tafenoquine Succinate E4946-100MG
SDP
Catalog No. 5000043354
Encompass_Preferred
Tafenoquine Succinate(WR 238605 Succinate Etaquine Krintafel) is an 8-aminoquinoline (8AQ) anti-malarial drug that kills both the liver and blood stages of the parasite broadening its applicability for chemoprophylaxis to all species of malaria It is specifically used for the prevention of Plasmodium vivax relapse
Catalog No. 50-000-43354 Supplier Selleck Chemical LLC Supplier No. E4946100MG
Tafenoquine Succinate(WR 238605 Succinate Etaquine Krintafel) is an 8-aminoquinoline (8AQ) anti-malarial drug that kills both the liver and blood stages of the parasite broadening its applicability for chemoprophylaxis to all species of malaria It is specifically used for the prevention of Plasmodium vivax relapse