Learn More
Cayman Chemical Tafenoqun 10mg
SDP
Catalog No. 502990051
Encompass_Preferred
An antimalarial agent; active against multidrug-resistant clinical isolates of P. falciparum (IC50s = 64.6-110.4 ng/ml); inhibits sporozoite transmission to mosquitos fed on P. berghei-infected mice at 25 or 50 mg/kg; suppresses parasitemia in a hamster model of B. microti infection from 3.25-52 mg/kg
Catalog No. 50-299-0051 Supplier Cayman Chemical Supplier No. 3067310 MG
An antimalarial agent; active against multidrug-resistant clinical isolates of P. falciparum (IC50s = 64.6-110.4 ng/ml); inhibits sporozoite transmission to mosquitos fed on P. berghei-infected mice at 25 or 50 mg/kg; suppresses parasitemia in a hamster model of B. microti infection from 3.25-52 mg/kg