Learn More
Haematologic Technologies Inc TAFI 50UG
Catalog No. 50883461
Encompass_Preferred
Inhibitor, Thrombin Activatable Fibrinolysis Inhibitor, 50 micrograms, Purified from plasma, Action: Inhibition of fibrinolysis by removal of plasminogen binding sites on fibrin, Research use only, Stored in Hepes buffered saline, Store at -80 C.
Catalog No. 50-883-461 Supplier Haematologic Technologies Inc Supplier No. TAFI01
Inhibitor, Thrombin Activatable Fibrinolysis Inhibitor, 50 micrograms, Purified from plasma, Action: Inhibition of fibrinolysis by removal of plasminogen binding sites on fibrin, Research use only, Stored in Hepes buffered saline, Store at -80 C.