Learn More
Selleck Chemical LLC Tafluprost S4851-2mg
SDP
Supplier:  Selleck Chemical LLC S48512MG
Encompass_Preferred
Tafluprost (AFP-168 MK2452) is a novel prostaglandin analog with a high affinity for the fluoroprostaglandin (FP) receptor PGF2 A prostanoid selective FP receptor agonist
Catalog No. 50-311-1390