Learn More
Cayman Chemical Tafluprost-d4 100UG
SDP
Catalog No. 502892748
Encompass_Preferred
An internal standard for the quantification of tafluprost by GC- or LC-MS
Catalog No. 50-289-2748 Supplier Cayman Chemical Supplier No. 9002405100 UG
An internal standard for the quantification of tafluprost by GC- or LC-MS