Learn More
Cayman Chemical Tafluprost-d4 500UG
SDP
Catalog No. 502893753
Encompass_Preferred
An internal standard for the quantification of tafluprost by GC- or LC-MS
Catalog No. 50-289-3753 Supplier Cayman Chemical Supplier No. 9002405500 UG
An internal standard for the quantification of tafluprost by GC- or LC-MS