Learn More
Cayman Chemical Tafluprost-d4 50UG
SDP
Catalog No. 502893658
Encompass_Preferred
An internal standard for the quantification of tafluprost by GC- or LC-MS
Catalog No. 50-289-3658 Supplier Cayman Chemical Supplier No. 900240550 UG
An internal standard for the quantification of tafluprost by GC- or LC-MS