Learn More
Medchemexpress LLC Tafluprost ethyl amide | 1185851-52-8 | 99.6% | 437.52 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY139420100MG
Encompass_Preferred
Tafluprost ethyl amide is a prostaglandin derivative. It is capable of reducing intraocular pressure (IOP) and influencing eyelash growth. It can be used in antiglaucoma ophthalmic compositions.
  • Capable of reducing intraocular pressure (IOP)
  • Influences eyelash growth
  • Used in antiglaucoma ophthalmic compositions
  • Acts as a prostaglandin receptor inhibitor
Catalog No. 50-003-51153