Learn More
Medchemexpress LLC Tafluprost ethyl amide | 1185851-52-8 | 99.6% | 437.52 | 25 MG
SDP
Catalog No. 5000349146
Encompass_Preferred
Tafluprost ethyl amide is a prostaglandin derivative that can reduce intraocular pressure (IOP) and influence eyelash growth. It can be utilized in antiglaucoma ophthalmic compositions.
  • Prostaglandin derivative
  • Reduces intraocular pressure (IOP)
  • Influences eyelash growth
  • Utilized in antiglaucoma ophthalmic compositions
Catalog No. 50-003-49146 Supplier Medchemexpress LLC Supplier No. 25MGHY139420
Tafluprost ethyl amide is a prostaglandin derivative that can reduce intraocular pressure (IOP) and influence eyelash growth. It can be utilized in antiglaucoma ophthalmic compositions.
  • Prostaglandin derivative
  • Reduces intraocular pressure (IOP)
  • Influences eyelash growth
  • Utilized in antiglaucoma ophthalmic compositions