Learn More
Medchemexpress LLC Tafluprost ethyl amide | 1185851-52-8 | 99.6% | C24H33F2NO4 | 10MG
SDP
Catalog No. 5000261656
Encompass_Preferred
Tafluprost ethyl amide is a prostaglandin derivative used in research to reduce intraocular pressure and to investigate effects on eyelash growth. Supplied for research use only, it is intended for pharmacological and preclinical ophthalmic studies involving prostaglandin receptor activity.
  • Reduces intraocular pressure in experimental settings.
  • Influences eyelash growth pathways for cosmetic and biological research.
  • High reported purity suitable for analytical and preclinical work.
  • Offered in multiple small-scale package sizes for laboratory use.
  • Stable under recommended storage conditions when protected from light.
Catalog No. 50-002-61656 Supplier Medchemexpress LLC Supplier No. HY13942010MG
Tafluprost ethyl amide is a prostaglandin derivative used in research to reduce intraocular pressure and to investigate effects on eyelash growth. Supplied for research use only, it is intended for pharmacological and preclinical ophthalmic studies involving prostaglandin receptor activity.
  • Reduces intraocular pressure in experimental settings.
  • Influences eyelash growth pathways for cosmetic and biological research.
  • High reported purity suitable for analytical and preclinical work.
  • Offered in multiple small-scale package sizes for laboratory use.
  • Stable under recommended storage conditions when protected from light.