Learn More
Cayman Chemical Tafluprost free acid
SDP
Catalog No. 502890921
Encompass_Preferred
An FP receptor agonist (Ki = 4 nM for the human receptor), a PGF2α derivative, and an active metabolite of tafluprost; selective for the FP receptor over the dopamine receptor and PGE2 receptor subtypes EP1 and EP2, as well as a panel of 32 neurological receptors and transporters, at 1 µM; induces constriction in isolated cat iris sphincters (EC50 = 0.6 nM); increases the proliferation and migration of, capillary formation by, and COX-2 levels in, HUVECs at 100 µM
Catalog No. 50-289-0921 Supplier Cayman Chemical Supplier No. 100054395 MG
Add to Cart
Add to Cart
An FP receptor agonist (Ki = 4 nM for the human receptor), a PGF2α derivative, and an active metabolite of tafluprost; selective for the FP receptor over the dopamine receptor and PGE2 receptor subtypes EP1 and EP2, as well as a panel of 32 neurological receptors and transporters, at 1 µM; induces constriction in isolated cat iris sphincters (EC50 = 0.6 nM); increases the proliferation and migration of, capillary formation by, and COX-2 levels in, HUVECs at 100 µM