Learn More
Cayman Chemical Tafluprostfree acId-d4 500UG
SDP
Catalog No. 502892583
Encompass_Preferred
An internal standard for the quantification of tafluprost (free acid) by GC-or LC-MS
Catalog No. 50-289-2583 Supplier Cayman Chemical Supplier No. 9002406500 UG
An internal standard for the quantification of tafluprost (free acid) by GC-or LC-MS