Learn More
Enzo Life Sciences Tumor associated glycoprotein TAG-72 Recombinant monoclonal antibody (B72.3) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5670200
Encompass_Preferred
Clone: B72.3 (Satumomab). Isotype: IgGκ. Immunogen: Human tumor associated glycoprotein 72. Species reactivity: Human. Applications: Flow Cytometry, IHC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3093