Learn More
Research Products International Corp TAG, Thermal, 0.81x0.28"", 4000/pk
SDP
Supplier:  Research Products International Corp 249430C3
Encompass
Thermal Transfer Cryo-Tags are designed for use with Zebra brand printers. Quickly print barcodes and text clearly for fast and easy sample identification. Cryogenic labels provides superb cold temperature performance from -196°C to +80°C and work in liquid/vapor phase applications and withstand flash/snap freezing in liquid nitrogen.
Labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, micro-tubes, test tubes, glass vials, beakers and metal racks. Durable material resists degradation from scuffing, chemicals, moisture and temperature fluctuations. Supplied in large assortment of shapes and sizes on 1” or 3” core.
-196°C to +80°C
Cryogenic, low temperature
Roll format
Catalog No. 50-222-9615