Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 182875MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: CC49. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Jurkat cells, Breast or lung carcinoma. Cellular Staining: Cytoplasmic, cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Purified TAG-72 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 220kDa.Chromosome: Not Known. Gene Symbol: Not Known. Gene ID: 182875. Ensemble: WBGene00006447. Uniprot: NP_492674.
Catalog No. 50-188-4646