Learn More
Aviva Systems Biology Corp TAGAP Antibody - N-terminal region (ARP62158_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62158_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TAGAP. It was validated on Western Blot. . Alternate names: FKSG15, FLJ32631, FLJ39771, IDDM21, MGC133247, MGC27381, TAGAP1, ARHGAP47ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-591