Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 6876MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: TAGLN/247. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: U-2 OS, Hs68 or WI 38 cells. Colon carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human transgelin (TAGLN) protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 22kDa.Chromosome: 11q23.2. Gene Symbol: TAGLN. Gene ID: 6876. Ensemble: ENSG00000149591. Uniprot: NP_003177.
Catalog No. 50-188-5196