Learn More
Aviva Systems Biology Corp Tagln3 Antibody - N-terminal region (ARP54395_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54395_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tagln3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2700038H05Rik, 2900005O10Rik, AI426007, Np25ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-921