Learn More
Apexbio Technology LLC TAI-1, 5mg. Cas: 1334921-03-7 MFCD: MFCD28348371. Hec1 inhibitor, potent, first-in-class
SDP
Supplier:  Apexbio Technology LLC B48925
Encompass_Preferred
TAI-1 is a highly potent, first-in-class inhibitor of Hec1 with IC50 of TAI-1 has a GI50 of 13.48 nM (K562 cells), which is close to 1000 times improvement in potency compared to INH1 (GI50 =11.7 M). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5362