Learn More
Braintree Scientific Tail Snipping Platform
SDP
Catalog No. 501954962
Encompass
Tail Snipping Platform For isolating DNA from transgenic animals for genotyping. Simply pull mouse tail through the slot, measure and collect tip in the vial. Features Micro Tube Holder: Collect tail tip directly in the prelabeled tube, preventing DNA cross contamination. Measuring Ruler (mm): Accurately measure tail length for cutting. Enhances IACUC compliance. Cleaning Slot: Feces and urine are easily washed out with stream of water (minimizes cross contamination). **Sold for research purposes only and not for human use**
Catalog No. 50-195-4962 Supplier Braintree Scientific Supplier No. IL200
 Restricted Item
Add to Cart
Add to Cart
Tail Snipping Platform For isolating DNA from transgenic animals for genotyping. Simply pull mouse tail through the slot, measure and collect tip in the vial. Features Micro Tube Holder: Collect tail tip directly in the prelabeled tube, preventing DNA cross contamination. Measuring Ruler (mm): Accurately measure tail length for cutting. Enhances IACUC compliance. Cleaning Slot: Feces and urine are easily washed out with stream of water (minimizes cross contamination). **Sold for research purposes only and not for human use**