Learn More
Stoelting™ Tail Suspension
SDP
Catalog No. 10000299 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse, Rat
Catalog No. Animal
10-000-299 Mouse, Rat
1 options
Catalog No. 10-000-299 Supplier Stoelting™ Supplier No. 60125
Add to Cart
Add to Cart
Stoelting™ Tail Suspension test is akin to the forced swim test but avoids the problems of hypothermia and stress associated with forced swimming.
Animals are suspended by their tails and the amount of immobility is measured. Longer periods of immobility are associated with higher depressive scores and immobility can be reversed with antidepressant treatment.
Specifications
For Use With (Application) Neuroscience
Animal Mouse, Rat