Learn More
SAI Infusion Technologies Rodent Tail Vein Catheter - 28cm, 1F tip, Stylet
SDP
Catalog No. NC9764401
Encompass
28cm PU Catheter with a 1F radiopaque tip, stylet, and female luer for easy syringe access. Sterile. The MTV-01 is SAI's best-selling tail vein catheter for mice and rats. It consists of a very small catheter and stylet that easily inserts into the tail vein of a restrained rodent. The size allows for quick, mistake free access to the tail vein and once inserted, the flexibility keeps vessel trauma to a minimum. Unlike sharp, stiff, needles this catheter moves freely up the vein to ensure your dose goes into the bloodstream, not into the surrounding tissue or back out through the insertion site. It bends naturally with the tail, reducing injury and allowing for continuous infusion or repeated bolus dosing with less chance of losing the tail. Great for short term access in restrained or anesthetized subjects. Product is for laboratory animal research only.
Catalog No. NC9764401 Supplier SAI Infusion Technologies Supplier No. MTV01
28cm PU Catheter with a 1F radiopaque tip, stylet, and female luer for easy syringe access. Sterile. The MTV-01 is SAI's best-selling tail vein catheter for mice and rats. It consists of a very small catheter and stylet that easily inserts into the tail vein of a restrained rodent. The size allows for quick, mistake free access to the tail vein and once inserted, the flexibility keeps vessel trauma to a minimum. Unlike sharp, stiff, needles this catheter moves freely up the vein to ensure your dose goes into the bloodstream, not into the surrounding tissue or back out through the insertion site. It bends naturally with the tail, reducing injury and allowing for continuous infusion or repeated bolus dosing with less chance of losing the tail. Great for short term access in restrained or anesthetized subjects. Product is for laboratory animal research only.