Learn More
MSC Meyer Products Tailgate Spreader, 350 lbs Capacity, Poly, Stainless Steel
Catalog No. 502836177
Encompass
This tailgate spreader efficiently spreads bagged rock salt. It features a high-density poly hopper with 350 lbs capacity and a stainless steel spinner, ensuring reliable and even material distribution.
  • Patented brush auger prevents ice melt leakage and aids material flow.
  • Powerful direct-drive motor provides superior auger power for consistent spreading.
  • Durable, rust-proof poly hopper includes heavy-duty locking straps.
  • Nine-inch stainless steel spinner offers robust and reliable performance.
  • Weatherproof electrical connections ensure protection against the elements.
  • One-touch auto control with illuminated in-cab controller for easy use.
  • No material waste feature automatically stops the spinner when braking.
  • Compatible with ½-ton and larger pickup trucks or tractors.
Catalog No. 50-283-6177 Supplier MSC Supplier No. 69483014
This tailgate spreader efficiently spreads bagged rock salt. It features a high-density poly hopper with 350 lbs capacity and a stainless steel spinner, ensuring reliable and even material distribution.
  • Patented brush auger prevents ice melt leakage and aids material flow.
  • Powerful direct-drive motor provides superior auger power for consistent spreading.
  • Durable, rust-proof poly hopper includes heavy-duty locking straps.
  • Nine-inch stainless steel spinner offers robust and reliable performance.
  • Weatherproof electrical connections ensure protection against the elements.
  • One-touch auto control with illuminated in-cab controller for easy use.
  • No material waste feature automatically stops the spinner when braking.
  • Compatible with ½-ton and larger pickup trucks or tractors.