Learn More
MSC Meyer Products Tailgate Spreader, 400 lb Capacity, 25 ft Spread Width, 1/8 HP Electric, Poly Hopper
Catalog No. 502837483
Encompass
The Base Line BL-400 Tailgate Spreader is designed for small- to medium-sized jobs, attaching to light trucks, SUVs, tractors, or utility vehicles with a 2 inch receiver hitch. It features a 400 lb capacity, 25 ft spread width, and a poly hopper for spreading bagged rock salt.
  • Patented brush auger prevents ice melt leakage
  • No material waste
  • Polyurethane hopper
  • Carbon steel frame
  • 9 inch poly spinner
  • 1/8 HP electric motor
  • Easy-to-use variable-speed controller
  • Internal filtering screen breaks up large clumps of material
  • Impeller spinner shaft adjusts spread pattern from 3-4 feet to 8-10 feet
  • Integrated storage stand and motor guard
  • Built-in material agitator
Catalog No. 50-283-7483 Supplier MSC Supplier No. 69483055
The Base Line BL-400 Tailgate Spreader is designed for small- to medium-sized jobs, attaching to light trucks, SUVs, tractors, or utility vehicles with a 2 inch receiver hitch. It features a 400 lb capacity, 25 ft spread width, and a poly hopper for spreading bagged rock salt.
  • Patented brush auger prevents ice melt leakage
  • No material waste
  • Polyurethane hopper
  • Carbon steel frame
  • 9 inch poly spinner
  • 1/8 HP electric motor
  • Easy-to-use variable-speed controller
  • Internal filtering screen breaks up large clumps of material
  • Impeller spinner shaft adjusts spread pattern from 3-4 feet to 8-10 feet
  • Integrated storage stand and motor guard
  • Built-in material agitator